Assay Details
Target Gene Details
Entrez Gene ID: | 18573 |
Gene Name: | phosphodiesterase 1A, calmodulin-dependent |
Gene Aliases: |
AI987702, AW125737 |
Location: |
Chr.2:79834453-80196518 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde1a | NM_001159582.1 | 16 | 3263 | NP_001153054.1 |
NM_016744.4 | 15 | 3058 | NP_058024.2 | |
AK043647.1 | 15 | 3059 | BAC31606.1 | |
AK134679.1 | 1 | 541 | ||
BC090628.1 | 16 | 3004 | AAH90628.1 |
More Information
Set Membership: |
Intragenic Exonic |