Assay Details
Target Gene Details
Entrez Gene ID: | 66811 |
Gene Name: | dual oxidase maturation factor 2 |
Gene Aliases: |
9030623N16Rik |
Location: |
Chr.2:122298900-122302885 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Duoxa2 | NM_025777.2 | 6 | 1023 | NP_080053.1 |
AK008816.1 | 6 | 1023 | BAB25910.1 | |
AK018569.1 | 6 | 1025 | BAB31281.1 | |
BC031111.1 | 6 | 1010 | AAH31111.1 |
More Information
Set Membership: |
Intragenic Exonic |