Assay Details
Target Gene Details
Entrez Gene ID: | 240832 |
Gene Name: | torsin A interacting protein 2 |
Gene Aliases: |
1110020D10Rik, 15kDa, A130072J07, AA103493, AW060462, AW610675, C77739, Ifrg15, Lull1 |
Location: |
Chr.1:156035664-156068861 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tor1aip2 | NM_172843.4 | 5 | 1499 | NP_766431.3 |
AK038024.1 | 5 | 1761 | BAC29923.1 | |
AK080959.1 | 3 | 778 | BAC38096.1 | |
BC066790.1 | 3 | 705 | AAH66790.1 | |
CK635751.1 | 1 | 339 |