Assay Details
Target Gene Details
Entrez Gene ID: | 26360 |
Gene Name: | angiopoietin-like 2 |
Gene Aliases: |
AI593246, AW260363, Arp2 |
Location: |
Chr.2:33215951-33247725 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Angptl2 | NM_011923.4 | 4 | 1678 | NP_036053.2 |
AF125176.1 | 3 | 1165 | AAD55358.1 | |
AK037265.1 | 4 | 1571 | BAC29780.1 | |
AK155464.1 | 4 | 1679 | BAE33276.1 | |
AK187902.1 | 2 | 244 | ||
AK205240.1 | 2 | 244 | ||
AK214337.1 | 2 | 244 | ||
BC138609.1 | 4 | 1484 | ||
BC138610.1 | 4 | 1484 |
Target Gene Details
Entrez Gene ID: | 241308 |
Gene Name: | Ral GEF with PH domain and SH3 binding motif 1 |
Gene Aliases: |
5830418G11Rik, AI853783, AI854138, RALGEF2, RALGPS1A, RalGEF 2, mKIAA0351 |
Location: |
Chr.2:33133419-33371511 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ralgps1 | NM_001290570.1 | NP_001277499.1 | ||
NM_001290572.1 | NP_001277501.1 | |||
NM_175211.5 | NP_780420.1 | |||
AK076767.1 | BAC36473.1 | |||
AK129123.1 | BAC97933.1 | |||
AK170248.1 | ||||
BC072656.1 | AAH72656.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |