Assay Details
Target Gene Details
Entrez Gene ID: | 14269 |
Gene Name: | formin binding protein 1 |
Gene Aliases: |
1110057E06Rik, 2210010H06Rik, FBP1, Fbp17 |
Location: |
Chr.2:31026206-31142027 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fnbp1 | NM_001038700.2 | 15 | 3634 | NP_001033789.1 |
NM_001177648.1 | 17 | 3832 | NP_001171119.1 | |
NM_001177649.1 | 15 | 3637 | NP_001171120.1 | |
NM_001177650.1 | 14 | 3484 | NP_001171121.1 | |
AK077936.1 | 1 | 1369 | ||
AK122308.1 | 17 | 3832 | BAC65590.1 | |
AK149169.1 | 1 | 1628 | BAE28755.1 | |
AK151666.1 | 1 | 1288 | ||
AK152998.1 | 1 | 1288 | ||
AK170497.1 | 15 | 3587 | BAE41836.1 | |
AK172100.1 | 15 | 3578 | BAE42825.1 |