Assay Details
Target Gene Details
Entrez Gene ID: | 18019 |
Gene Name: | nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2 |
Gene Aliases: |
AI607462, NF-ATc2, NF-ATp, NFAT1, NFAT1-D, Nfatp |
Location: |
Chr.2:168476410-168601657 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nfatc2 | NM_001136073.2 | 9 | 2786 | NP_001129545.1 |
NM_001291168.1 | 9 | 2671 | NP_001278097.1 | |
NM_001291169.1 | 9 | 2671 | NP_001278098.1 | |
NM_001291170.1 | 9 | 2151 | NP_001278099.1 | |
NM_001291171.1 | 8 | 1635 | NP_001278100.1 | |
NM_001291173.1 | 8 | 1635 | NP_001278102.1 | |
NM_001291174.1 | 9 | 2151 | NP_001278103.1 | |
NM_001291175.1 | 9 | 2266 | NP_001278104.1 | |
NM_001291176.1 | 8 | 1750 | NP_001278105.1 | |
NM_001291177.1 | 9 | 2266 | NP_001278106.1 | |
NM_001291178.1 | 8 | 1750 | NP_001278107.1 | |
NM_010899.3 | 9 | 2786 | NP_035029.2 | |
AK161174.1 | 7 | 1361 | BAE36226.1 | |
EU887581.1 | 9 | 2552 | ||
EU887582.1 | 9 | 2612 | ||
EU887583.1 | 9 | 2149 | ||
EU887584.1 | 9 | 2249 | ||
EU887585.1 | 8 | 1633 | ||
EU887586.1 | 8 | 1733 | ||
EU887587.1 | 9 | 2552 | ||
EU887588.1 | 9 | 2612 | ||
EU887589.1 | 9 | 2149 | ||
EU887590.1 | 9 | 2249 | ||
EU887591.1 | 8 | 1633 | ||
EU887592.1 | 8 | 1733 | ||
U36575.1 | 9 | 2682 | AAC52930.1 | |
U36576.1 | 9 | 2682 | AAC52931.1 |
More Information
Set Membership: |
Intragenic Exonic |