Assay Details
Target Gene Details
Entrez Gene ID: | 227743 |
Gene Name: | mitogen-activated protein kinase associated protein 1 |
Gene Aliases: |
AI591529, D230039K05Rik, Sin1 |
Location: |
Chr.2:34406771-34624958 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mapkap1 | NM_001290625.1 | 12 | 2861 | NP_001277554.1 |
NM_001290626.1 | 11 | 2859 | NP_001277555.1 | |
NM_177345.4 | 12 | 3187 | NP_796319.1 | |
AK132263.1 | 12 | 2851 | BAE21066.1 | |
BC048870.1 | 1 | 458 | AAH48870.1 | |
BC090644.1 | 11 | 2761 | AAH90644.1 |
More Information
Set Membership: |
Intragenic Exonic |