Assay Details
Target Gene Details
Entrez Gene ID: | 18019 |
Gene Name: | nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2 |
Gene Aliases: |
AI607462, NF-ATc2, NF-ATp, NFAT1, NFAT1-D, Nfatp |
Location: |
Chr.2:168476410-168601657 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nfatc2 | NM_001037177.2 | 7 | 3159 | NP_001032254.1 |
NM_001037178.2 | 7 | 2524 | NP_001032255.1 | |
NM_001136073.2 | NP_001129545.1 | |||
NM_001291168.1 | NP_001278097.1 | |||
NM_001291169.1 | NP_001278098.1 | |||
NM_001291170.1 | NP_001278099.1 | |||
NM_001291171.1 | NP_001278100.1 | |||
NM_001291172.1 | 7 | 3044 | NP_001278101.1 | |
NM_001291173.1 | NP_001278102.1 | |||
NM_001291174.1 | NP_001278103.1 | |||
NM_001291175.1 | NP_001278104.1 | |||
NM_001291176.1 | NP_001278105.1 | |||
NM_001291177.1 | NP_001278106.1 | |||
NM_001291178.1 | NP_001278107.1 | |||
NM_001291179.1 | 7 | 2639 | NP_001278108.1 | |
NM_010899.3 | NP_035029.2 | |||
NR_111897.1 | 6 | 2008 | ||
NR_111898.1 | 6 | 2123 | ||
AK083119.1 | 7 | 2500 | BAC38768.1 | |
AK161174.1 | BAE36226.1 | |||
AW049827.1 | 1 | 240 | ||
EU887581.1 | ||||
EU887582.1 | ||||
EU887583.1 | ||||
EU887584.1 | ||||
EU887585.1 | ||||
EU887586.1 | ||||
EU887587.1 | ||||
EU887588.1 | ||||
EU887589.1 | ||||
EU887590.1 | ||||
EU887591.1 | ||||
EU887592.1 | ||||
EU887599.1 | 7 | 2925 | ||
EU887600.1 | 7 | 2985 | ||
EU887601.1 | 7 | 2522 | ||
EU887602.1 | 7 | 2622 | ||
EU887603.1 | 6 | 2006 | ||
EU887604.1 | 6 | 2106 | ||
U36575.1 | AAC52930.1 | |||
U36576.1 | AAC52931.1 |
More Information
Set Membership: |
Intragenic Exonic |