Assay Details
Target Gene Details
Entrez Gene ID: | 56406 |
Gene Name: | nuclear receptor coactivator 6 |
Gene Aliases: |
AIB3, ASC-2, ASC2, NRC, Ncoa7, PRIP, RAP250, mKIAA0181 |
Location: |
Chr.2:155390656-155473894 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncoa6 | NM_001242558.1 | 8 | 2581 | NP_001229487.1 |
NM_019825.3 | 8 | 2581 | NP_062799.2 | |
AF135169.1 | 1 | 79 | AAF35973.1 | |
AF216186.1 | 7 | 2466 | AAF35860.1 | |
AK142566.1 | 2 | 766 | BAE25107.1 | |
AK144338.1 | 10 | 2819 | ||
AK171864.1 | 8 | 2739 | BAE42705.1 | |
BC083187.1 | 8 | 2581 | AAH83187.1 | |
JF707634.1 | 7 | 2440 | ||
JF707635.1 | 7 | 2440 | ||
JF707636.1 | 7 | 2440 |
More Information
Set Membership: |
Intragenic Exonic |