Assay Details
Target Gene Details
Entrez Gene ID: | 74104 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 6 |
Gene Aliases: |
1200005B17Rik |
Location: |
Chr.1:75171640-75180283 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abcb6 | NM_023732.2 | 19 | 2758 | NP_076221.1 |
AK004605.1 | BAB23404.1 | |||
AK151165.1 | 19 | 2867 | BAE30168.1 | |
AK164003.1 | 16 | 3676 | ||
AK168642.1 | 19 | 2859 | BAE40501.1 | |
AK195614.1 | 3 | 282 | ||
AK211025.1 | 3 | 269 | ||
AK217592.1 | 3 | 269 | ||
BC006634.1 | 19 | 2758 | AAH06634.1 |
More Information
Set Membership: |
Intragenic Exonic |