Assay Details
Target Gene Details
Entrez Gene ID: | 116914 |
Gene Name: | solute carrier family 19 (thiamine transporter), member 2 |
Gene Aliases: |
AV276020, AW322295, DDA1, THTR1, TRMA |
Location: |
Chr.1:164249046-164265385 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc19a2 | NM_001276455.1 | 2 | 609 | NP_001263384.1 |
NM_054087.3 | 2 | 609 | NP_473428.1 | |
AF179403.1 | 2 | 600 | AAG43424.1 | |
AF216204.1 | 2 | 579 | AAK33067.1 | |
AF326916.1 | 2 | 402 | AAL85635.1 | |
AF360396.1 | 2 | 591 | AAL30451.1 | |
AF418986.1 | 2 | 600 | AAM47550.1 | |
AK044825.1 | 2 | 610 | BAC32108.1 | |
BC034659.1 | 2 | 590 | AAH34659.1 |
More Information
Set Membership: |
Intragenic Exonic |