Assay Details
Target Gene Details
Entrez Gene ID: | 227157 |
Gene Name: | membrane protein, palmitoylated 4 (MAGUK p55 subfamily member 4) |
Gene Aliases: |
Dlg6 |
Location: |
Chr.1:59120935-59163389 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpp4 | NM_001164682.1 | 23 | 2732 | NP_001158154.1 |
NM_145143.3 | 22 | 2095 | NP_660125.2 | |
AB059357.1 | 21 | 1986 | BAB69494.1 | |
AB059358.1 | 20 | 1968 | BAB69495.1 | |
AK044682.1 | 23 | 2731 | BAC32031.1 | |
BC061694.1 | 22 | 2012 | ||
BC116723.1 | 17 | 1615 |
More Information
Set Membership: |
Intragenic Exonic |