Assay Details
Target Gene Details
Entrez Gene ID: | 67769 |
Gene Name: | G patch domain containing 2 |
Gene Aliases: |
5830433G22Rik, 5830436K05Rik, AI427714, AI447508, AW107440, AW491060, Gpatc2 |
Location: |
Chr.1:187215393-187369480 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpatch2 | NM_026367.4 | 10 | 1988 | NP_080643.4 |
AK029990.1 | 10 | 1972 | ||
AK030026.1 | 10 | 1987 | BAC26744.1 | |
AK053781.1 | 8 | 1869 | BAC35520.1 | |
AK083471.1 | 9 | 1145 | BAC38928.1 | |
AK169660.1 | 11 | 2109 | ||
BC054810.1 | 10 | 1960 | AAH54810.1 |
More Information
Set Membership: |
Intragenic Exonic |