Assay Details
Target Gene Details
Entrez Gene ID: | 14262 |
Gene Name: | flavin containing monooxygenase 3 |
Gene Aliases: |
AW111792 |
Location: |
Chr.1:162953800-162984491 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fmo3 | NM_008030.1 | 9 | 1654 | NP_032056.1 |
AK149440.1 | 9 | 1681 | BAE28877.1 | |
U87147.1 | 9 | 1654 | AAB47541.1 |
More Information
Set Membership: |
Intragenic Exonic |