Assay Details
Target Gene Details
Entrez Gene ID: | 241134 |
Gene Name: | neuronal tyrosine-phophorylated phosphoinositide 3-kinase adaptor 2 |
Gene Aliases: |
9430031J16Rik, Jr6, Kiaa1486 |
Location: |
Chr.1:81076553-81365845 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nyap2 | NM_172849.3 | 3 | 883 | NP_766437.2 |
AB429290.1 | 3 | 374 | ||
AK034747.1 | 4 | 898 | BAC28819.1 | |
AK043512.1 | 3 | 618 | ||
AK049778.1 | 4 | 892 | BAC33918.1 | |
AK137094.1 | 3 | 880 | BAE23236.1 | |
BC082310.1 | 3 | 883 | AAH82310.1 |
More Information
Set Membership: |
Intragenic Exonic |