Assay Details
Target Gene Details
Entrez Gene ID: | 108954 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 15b |
Gene Aliases: |
1810033K10Rik, AI606441, C530022L24Rik, CReP |
Location: |
Chr.1:133131166-133139800 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r15b | NM_133819.3 | 1 | 1015 | NP_598580.1 |
AK049028.1 | 1 | 719 | BAC33517.1 | |
AK082957.1 | 1 | 851 | BAC38708.1 | |
AK086606.1 | 1 | 647 | BAC39701.1 | |
AK144678.1 | 1 | 768 | BAE26007.1 | |
AK146089.1 | 1 | 753 | BAE26892.1 | |
AK167565.1 | 1 | 838 | BAE39628.1 | |
AK170862.1 | 1 | 845 | BAE42079.1 | |
BC006897.1 | AAH06897.1 |