Assay Details
Target Gene Details
Entrez Gene ID: | 56695 |
Gene Name: | paroxysmal nonkinesiogenic dyskinesia |
Gene Aliases: |
2210013N15Rik, 2810403H05Rik, AI854243, Brp17, MNCb-5687, MR-1, Tahccp2 |
Location: |
Chr.1:74284597-74353692 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pnkd | NM_001039509.1 | 10 | 1988 | NP_001034598.1 |
NM_019999.2 | 9 | 1897 | NP_064383.1 | |
AB041609.1 | 9 | 1898 | BAA95092.1 | |
AK008723.1 | 1 | 458 | ||
AK173125.1 | 10 | 1962 | BAD32403.1 | |
AK183859.1 | ||||
BC058945.1 | 11 | 2084 | AAH58945.1 |