Assay Details
Target Gene Details
Entrez Gene ID: | 58181 |
Gene Name: | interleukin 20 |
Gene Aliases: |
Zcyto10 |
Location: |
Chr.1:130907192-130911296 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il20 | NM_021380.1 | 5 | 628 | NP_067355.1 |
AF224267.1 | 5 | 628 | AAF36680.1 | |
AK078698.1 | 5 | 785 | ||
AY876923.1 | AAW78349.1 | |||
BC111821.1 | 5 | 618 | AAI11822.1 |