Assay Details
Target Gene Details
Entrez Gene ID: | 226438 |
Gene Name: | immunoglobulin-like and fibronectin type III domain containing 1 |
Gene Aliases: |
9830123M21Rik |
Location: |
Chr.1:135953205-136040842 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igfn1 | NM_177642.3 | 27 | 8625 | NP_808310.2 |
AK036509.1 | 8 | 1400 | BAC29455.1 | |
BC107027.1 | AAI07028.1 | |||
BC107028.2 | AAI07029.1 | |||
EU429481.1 | 13 | 5497 | ||
EU429482.1 | 12 | 2364 | ||
GU290042.1 | 27 | 8625 |