Assay Details
Target Gene Details
Entrez Gene ID: | 12767 |
Gene Name: | chemokine (C-X-C motif) receptor 4 |
Gene Aliases: |
CD184, Cmkar4, LESTR, PB-CKR, PBSF/SDF-1, Sdf1r, b2b220Clo |
Location: |
Chr.1:128588199-128592315 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcr4 | NM_009911.3 | 2 | 1534 | NP_034041.2 |
AB000803.1 | 2 | 1515 | BAA19187.1 | |
AK088663.1 | 1 | 248 | ||
BC031665.1 | 2 | 1521 | AAH31665.1 | |
BC098322.1 | 2 | 1511 | AAH98322.1 | |
BF662224.1 | 1 | 254 | ||
X99582.1 | CAA67894.1 |