Assay Details
Target Gene Details
Entrez Gene ID: | 58175 |
Gene Name: | regulator of G-protein signaling 20 |
Gene Aliases: |
2900073E09Rik, Rgsz1 |
Location: |
Chr.1:4909573-5070308 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs20 | NM_001177795.1 | 6 | 1622 | NP_001171266.1 |
NM_001290372.1 | 5 | 1205 | NP_001277301.1 | |
NM_021374.5 | 5 | 1275 | NP_067349.2 | |
AK013773.1 | BAB28987.2 | |||
AK078208.1 | 5 | 1120 | BAC37173.1 | |
AK089947.1 | 5 | 1095 | ||
AK158864.1 | 5 | 1116 | BAE34700.1 |
More Information
Set Membership: |
Intragenic Exonic |