Assay Details
Target Gene Details
Entrez Gene ID: | 18703 |
Gene Name: | polymeric immunoglobulin receptor |
Gene Aliases: |
- |
Location: |
Chr.1:130826684-130852249 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pigr | NM_011082.3 | 5 | 1388 | NP_035212.2 |
AK128944.1 | 5 | 1388 | ||
AK142656.1 | 5 | 1390 | BAE25149.1 | |
BC013556.1 | 5 | 1321 | AAH13556.1 | |
U06431.1 | 5 | 1292 | AAA67440.1 |
More Information
Set Membership: |
Intragenic Exonic |