Assay Details
Target Gene Details
Entrez Gene ID: | 329252 |
Gene Name: | leucine-rich repeat-containing G protein-coupled receptor 6 |
Gene Aliases: |
A530037C04Rik, D830026M09 |
Location: |
Chr.1:134983300-135105374 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lgr6 | NM_001033409.3 | 18 | 3012 | NP_001028581.1 |
AK040883.1 | 1 | 613 | ||
AK052873.1 | 13 | 2212 | ||
AK085901.1 | 18 | 2971 | ||
AK137380.1 | 18 | 3013 | BAE23335.1 | |
BC026896.1 | 3 | 1354 | AAH26896.1 | |
BC141210.1 | 18 | 2964 |