Assay Details
Target Gene Details
Entrez Gene ID: | 100503816 |
Gene Name: | predicted gene 16548 |
Gene Aliases: |
- |
Location: |
Chr.1:164148576-164152217 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm16548 | NR_037987.1 | 1 | 223 | |
AK149537.1 |