Assay Details
Target Gene Details
Entrez Gene ID: | 67426 |
Gene Name: | aarF domain containing kinase 3 |
Gene Aliases: |
4632432J16Rik, AI462003, Cabc1, mKIAA0451 |
Location: |
Chr.1:180165238-180196020 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adck3 | NM_001163290.1 | 15 | 3184 | NP_001156762.1 |
NM_023341.3 | 15 | 3282 | NP_075830.2 | |
AK014605.2 | 15 | 3224 | BAB29459.2 |