Assay Details
Target Gene Details
Entrez Gene ID: | 12283 |
Gene Name: | calcium binding protein 39 |
Gene Aliases: |
AA408805, AA960512, C78372, MO25, MO25alpha |
Location: |
Chr.1:85793410-85851577 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cab39 | NM_133781.4 | 9 | 3375 | NP_598542.3 |
AK003017.1 | 1 | 992 | ||
AK004036.1 | 1 | 980 | ||
AK005226.1 | 1 | 1302 | ||
AK144665.1 | 10 | 3208 | BAE25997.1 | |
BB694495.1 | 1 | 119 | ||
BC029053.1 | 5 | 2549 | AAH29053.1 |