Assay Details
Target Gene Details
Entrez Gene ID: | 319263 |
Gene Name: | protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1 |
Gene Aliases: |
8430411F12Rik, A030012M09Rik |
Location: |
Chr.1:7088891-7173628 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcmtd1 | NM_183028.3 | 6 | 4535 | NP_898849.1 |
AK018402.1 | 1 | 390 | ||
BC110360.1 | 6 | 4305 | AAI10361.1 |
More Information
Set Membership: |
Intragenic Exonic |