Assay Details
Target Gene Details
Entrez Gene ID: | 51801 |
Gene Name: | receptor (calcitonin) activity modifying protein 1 |
Gene Aliases: |
9130218E19Rik |
Location: |
Chr.1:91179822-91225196 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ramp1 | NM_016894.3 | 3 | 1502 | NP_058590.1 |
AK020282.1 | 1 | 176 | ||
AK165238.1 | 3 | 1278 | BAE38095.1 |
More Information
Set Membership: |
Intragenic Exonic |