Assay Details
Target Gene Details
Entrez Gene ID: | 26900 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked |
Gene Aliases: |
8030469F12Rik, D1Pas1-rs1, Dby |
Location: |
Chr.Y:1260715-1286628 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx3y | NM_012008.2 | 16 | 1933 | NP_036138.1 |
AJ007376.1 | 16 | 1900 | CAA07483.1 | |
AK154382.1 | 16 | 1893 | BAE32549.1 | |
AK160604.1 | 3 | 422 | BAE35907.1 | |
BC021453.1 | 16 | 1898 | AAH21453.1 |