Assay Details
Target Gene Details
Entrez Gene ID: | 93761 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1 |
Gene Aliases: |
5730494M04Rik, Snf2l |
Location: |
Chr.X:47809370-47892931 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smarca1 | NM_001290708.1 | 23 | 3230 | NP_001277637.1 |
NM_053123.4 | 23 | 3230 | NP_444353.3 | |
AF325920.1 | 23 | 3240 | AAK52453.1 | |
AK030741.1 | 23 | 3197 | BAC27109.1 | |
BC016071.1 | ||||
BC057115.1 | 23 | 3230 | AAH57115.1 |
More Information
Set Membership: |
Intragenic Exonic |