Assay Details
Target Gene Details
Entrez Gene ID: | 75974 |
Gene Name: | dedicator of cytokinesis 11 |
Gene Aliases: |
5033414A21Rik, 8030476J24, Zizimin2, sph238 |
Location: |
Chr.X:35888627-36076570 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dock11 | NM_001009947.3 | 6 | 611 | NP_001009947.2 |
AB116935.2 | 6 | 611 | BAD83670.1 |
More Information
Set Membership: |
Intragenic Exonic |