Assay Details
Target Gene Details
Entrez Gene ID: | 14381 |
Gene Name: | glucose-6-phosphate dehydrogenase X-linked |
Gene Aliases: |
G28A, G6pd, Gpdx |
Location: |
Chr.X:74409486-74429161 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
G6pdx | NM_008062.2 | NP_032088.1 | ||
AK088135.1 | BAC40166.1 | |||
AK148031.1 | ||||
AK165207.1 | BAE38077.1 | |||
AK183482.1 | ||||
BC075663.1 | AAH75663.1 | |||
Z11911.1 | CAA77967.1 |
Target Gene Details
Entrez Gene ID: | 16151 |
Gene Name: | inhibitor of kappaB kinase gamma |
Gene Aliases: |
1110037D23Rik, AI848108, AI851264, AW124339, IKK[g], NEMO |
Location: |
Chr.X:74393291-74456792 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ikbkg | NM_001161423.1 | NP_001154895.1 | ||
NM_001161424.1 | NP_001154896.1 | |||
CJ099503.1 | ||||
CN717929.1 | ||||
CV556906.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |