Assay Details
Target Gene Details
Entrez Gene ID: | 53412 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 3C |
Gene Aliases: |
PTG, Ppp1r5 |
Location: |
Chr.19:36731731-36736604 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r3c | NM_016854.2 | NP_058550.1 | ||
AK078735.1 | ||||
BC052769.1 | AAH52769.1 | |||
BC054739.1 | AAH54739.1 |
More Information
Set Membership: |
Intragenic Non-exonic |