Assay Details
Target Gene Details
Entrez Gene ID: | 18719 |
Gene Name: | phosphatidylinositol-4-phosphate 5-kinase, type 1 beta |
Gene Aliases: |
PI4P5K-I[b], PIP5K1-beta, PIP5KIbeta, Pipk5b, STM7 |
Location: |
Chr.19:24294794-24555827 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pip5k1b | NM_008846.2 | 13 | 1581 | NP_032872.1 |
AF048695.1 | 11 | 1430 | AAC05374.1 | |
AK133809.1 | 13 | 1576 | ||
AK146100.1 | 13 | 1581 | BAE26901.1 | |
AK149354.1 | 13 | 1581 | BAE28830.1 | |
AK164552.1 | 13 | 1564 | BAE37836.1 | |
BC034864.1 | 12 | 1439 | AAH34864.1 | |
D86176.1 | 12 | 1514 | BAA13030.1 |
More Information
Set Membership: |
Intragenic Exonic |