Assay Details
Target Gene Details
Entrez Gene ID: | 94067 |
Gene Name: | mitochondrial ribosomal protein L43 |
Gene Aliases: |
4930442D21Rik, bMRP36a |
Location: |
Chr.19:45005014-45006442 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrpl43 | NM_053164.3 | NP_444394.1 | ||
AB049657.1 | BAB40862.1 | |||
AK015357.1 | ||||
AK076204.1 | ||||
BC027676.1 | AAH27676.1 | |||
BC048733.1 | AAH48733.1 | |||
BC065812.1 | AAH65812.1 | |||
BY251956.1 |
Target Gene Details
Entrez Gene ID: | 226153 |
Gene Name: | progressive external ophthalmoplegia 1 (human) |
Gene Aliases: |
D19Ertd626e, PEO, Twinl |
Location: |
Chr.19:45006079-45012762 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Peo1 | AK054159.1 | 1 | 84 | BAE20696.1 |
More Information
Set Membership: |
Intragenic Non-exonic |