Assay Details
Target Gene Details
Entrez Gene ID: | 11668 |
Gene Name: | aldehyde dehydrogenase family 1, subfamily A1 |
Gene Aliases: |
Ahd-2, Ahd2, Aldh1, Aldh1a2, E1, Raldh1 |
Location: |
Chr.19:20601982-20643462 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aldh1a1 | NM_013467.3 | NP_038495.2 | ||
BC044729.2 | AAH44729.1 | |||
BC054386.1 | AAH54386.1 | |||
M74570.1 | AAA37202.1 | |||
S75713.1 | AAB32754.2 |
More Information
Set Membership: |
Intragenic Non-exonic |