Assay Details
Target Gene Details
Entrez Gene ID: | 12460 |
Gene Name: | copper chaperone for superoxide dismutase |
Gene Aliases: |
Ccsd |
Location: |
Chr.19:4825366-4839327 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccs | NM_016892.3 | NP_058588.1 | ||
AF121906.1 | AAD23832.1 | |||
AF173379.1 | AAF70242.1 | |||
AK010264.1 | BAB26806.1 | |||
AK050072.1 | BAC34057.1 | |||
AK192205.1 | ||||
BC026938.1 | AAH26938.1 |
More Information
Set Membership: |
Intragenic Non-exonic |