Assay Details
Target Gene Details
Entrez Gene ID: | 67160 |
Gene Name: | eukaryotic translation elongation factor 1 gamma |
Gene Aliases: |
2610301D06Rik, AA407312, EF1G |
Location: |
Chr.19:8967041-8978180 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eef1g | NM_026007.4 | 1 | 51 | NP_080283.3 |
AK145519.1 | 1 | 52 | BAE26482.1 |