Assay Details
Target Gene Details
Entrez Gene ID: | 13095 |
Gene Name: | cytochrome P450, family 2, subfamily c, polypeptide 29 |
Gene Aliases: |
AHOH, AHOHase, AI159681, AI196010, Ah-2, Ahh-1, Cyp2c, P450-2C |
Location: |
Chr.19:39287044-39330713 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp2c29 | NM_007815.3 | 1 | 50 | NP_031841.3 |
AI182545.1 | 1 | 61 | ||
BC019908.1 | 1 | 32 | AAH19908.1 | |
D17674.1 | BAA04555.1 |
More Information
Set Membership: |
Intragenic Exonic |