Assay Details
Target Gene Details
Entrez Gene ID: | 225912 |
Gene Name: | cytochrome b561 family, member A3 |
Gene Aliases: |
2310004G04Rik, Cybasc3, Lcytb |
Location: |
Chr.19:10577157-10595287 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyb561a3 | NM_001282064.1 | NP_001268993.1 | ||
NM_001282065.1 | NP_001268994.1 | |||
NM_001282067.1 | NP_001268996.1 | |||
NM_201351.2 | NP_958739.1 | |||
AK139533.1 | BAE24053.1 | |||
AK144666.1 | ||||
AK160179.1 | BAE35677.1 | |||
AK168841.1 | BAE40664.1 | |||
AK169729.1 | BAE41333.1 | |||
AK170911.1 | BAE42109.1 | |||
AK172638.1 | BAE43109.1 | |||
BC065078.1 | AAH65078.1 |
More Information
Set Membership: |
Intragenic Non-exonic |