Assay Details
Target Gene Details
Entrez Gene ID: | 433178 |
Gene Name: | serine peptidase inhibitor, Kazal type 14 |
Gene Aliases: |
EG433178, Gm5505 |
Location: |
Chr.18:44027869-44032208 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spink14 | NM_001039218.2 | 6 | 469 | NP_001034307.2 |
AK036889.1 | 1 | 3651 | BAC29624.1 | |
AV381433.2 | 5 | 576 | ||
BC147448.1 | ||||
BC147476.1 |
More Information
Set Membership: |
Intragenic Exonic |