Assay Details
Target Gene Details
Entrez Gene ID: | 16769 |
Gene Name: | desmoglein 4 |
Gene Aliases: |
CDHF13, lah |
Location: |
Chr.18:20436175-20471821 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dsg4 | NM_181564.2 | NP_853543.1 | ||
AB240192.1 | ||||
AY191584.1 | AAO91793.1 | |||
AY227349.1 | AAP44999.1 |
More Information
Set Membership: |
Intragenic Non-exonic |