Assay Details
Target Gene Details
Entrez Gene ID: | 225600 |
Gene Name: | phosphodiesterase 6A, cGMP-specific, rod, alpha |
Gene Aliases: |
Pdea, nmf282 |
Location: |
Chr.18:61220080-61289750 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde6a | NM_146086.2 | NP_666198.1 | ||
AK042142.1 | BAC31183.1 | |||
BC031925.1 | AAH31925.1 | |||
BC044892.1 | AAH44892.1 | |||
BC158126.1 | ||||
X60664.1 | CAA43072.1 |
More Information
Set Membership: |
Intragenic Non-exonic |