Assay Details
Target Gene Details
Entrez Gene ID: | 225600 |
Gene Name: | phosphodiesterase 6A, cGMP-specific, rod, alpha |
Gene Aliases: |
Pdea, nmf282 |
Location: |
Chr.18:61220080-61289750 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde6a | NM_146086.2 | 15 | 1991 | NP_666198.1 |
BC031925.1 | 15 | 1991 | AAH31925.1 | |
BC044892.1 | 15 | 1991 | AAH44892.1 | |
BC158126.1 | 15 | 2175 | ||
X60664.1 | 15 | 1999 | CAA43072.1 |
More Information
Set Membership: |
Intragenic Exonic |