Assay Details
Target Gene Details
Entrez Gene ID: | 56736 |
Gene Name: | ring finger protein 14 |
Gene Aliases: |
2310075C09Rik, 2610005D23Rik, AA986456, AU041447, D18Ertd188e, D7Bwg0165e, Triad2 |
Location: |
Chr.18:38296635-38317852 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rnf14 | NM_001164622.1 | 1 | 109 | NP_001158094.1 |
NM_020012.2 | 1 | 109 | NP_064396.2 | |
AF249667.1 | AAF74266.1 | |||
AK009783.1 | BAB26503.1 | |||
AK010162.1 | BAB26741.1 | |||
AK011316.2 | BAB27541.1 | |||
AK038986.1 | ||||
BC054841.1 | AAH54841.1 | |||
BC094250.1 | AAH94250.1 | |||
CA315275.1 | 1 | 109 |
More Information
Set Membership: |
Intragenic Exonic |