Assay Details
Target Gene Details
Entrez Gene ID: | 17991 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2 |
Gene Aliases: |
AV000592, B8, C1-B8, CI-B8 |
Location: |
Chr.18:36742332-36744587 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufa2 | NM_010885.5 | NP_035015.2 | ||
AF124786.1 | AAD30475.1 | |||
AK003608.1 | BAB22887.1 | |||
AK018732.1 | BAB31375.1 | |||
AV482131.1 | ||||
BC006815.1 | AAH06815.1 |
More Information
Set Membership: |
Intragenic Non-exonic |