Assay Details
Target Gene Details
Entrez Gene ID: | 18140 |
Gene Name: | ubiquitin-like, containing PHD and RING finger domains, 1 |
Gene Aliases: |
AL022808, ICBP90, Np95, RNF106 |
Location: |
Chr.17:56303337-56323486 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uhrf1 | NM_001111078.1 | NP_001104548.1 | ||
NM_001111079.1 | NP_001104549.1 | |||
NM_001111080.1 | NP_001104550.1 | |||
NM_010931.3 | NP_035061.3 | |||
AF274046.1 | AAK55743.1 | |||
AK143688.1 | BAE25499.1 | |||
AK145376.1 | BAE26398.1 | |||
AK145419.1 | ||||
AK145543.1 | BAE26496.1 | |||
AK145583.1 | ||||
AK146951.1 | BAE27560.1 | |||
AK147046.1 | BAE27632.1 | |||
AK150489.1 | BAE29605.1 | |||
AK151701.1 | BAE30624.1 | |||
AK151837.1 | BAE30730.1 | |||
AK152930.1 | BAE31605.1 | |||
AK153083.1 | BAE31708.1 | |||
BC022167.1 | AAH22167.1 | |||
D87908.1 | BAA74579.1 |
More Information
Set Membership: |
Intragenic Non-exonic |