Assay Details
Target Gene Details
Entrez Gene ID: | 319888 |
Gene Name: | O-acyltransferase like |
Gene Aliases: |
5330437I02Rik, Acyl3 |
Location: |
Chr.18:65697370-65751624 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Oacyl | NM_177028.3 | NP_796002.1 | ||
AK030604.1 | BAC27041.1 | |||
AK030615.1 | BAC27048.1 | |||
BC141265.1 |
More Information
Set Membership: |
Intragenic Non-exonic |