Assay Details
Target Gene Details
Entrez Gene ID: | 21354 |
Gene Name: | transporter 1, ATP-binding cassette, sub-family B (MDR/TAP) |
Gene Aliases: |
ABC17, APT1, Abcb2, Ham-1, Ham1, MTP1, PSF1, RING4, TAP, Tap-1, Y3 |
Location: |
Chr.17:34187556-34197225 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tap1 | NM_001161730.1 | NP_001155202.1 | ||
NM_013683.2 | NP_038711.2 | |||
AK040629.1 | ||||
AK040919.1 | 2 | 1033 | ||
AK151481.1 | ||||
AK152953.1 | BAE31620.1 | |||
AK153890.1 | BAE32237.1 | |||
AK162731.1 | BAE37041.1 | |||
AK166046.1 | ||||
AK167485.1 | ||||
AK171363.1 | BAE42411.1 | |||
BC013802.1 | AAH13802.1 | |||
BC024897.1 | AAH24897.1 | |||
KJ650227.1 | ||||
M55637.1 | AAA39570.1 | |||
U60018.1 | AAB41962.1 | |||
U60019.1 | AAB41963.1 | |||
U60020.1 | AAB41964.1 | |||
U60021.1 | AAB41965.1 | |||
U60022.1 | AAB41966.1 | |||
U60023.1 | AAB41967.1 | |||
X59615.1 | CAA42178.1 |
More Information
Set Membership: |
Intragenic Non-exonic |