Assay Details
Target Gene Details
Entrez Gene ID: | 269033 |
Gene Name: | RIKEN cDNA 4930503L19 gene |
Gene Aliases: |
9930107F24, BB148262, Lars, Las2 |
Location: |
Chr.18:70452210-70472504 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4930503L19Rik | NM_172967.2 | NP_766555.1 | ||
AK037066.1 | BAC29691.1 | |||
AK080780.1 | BAC38019.1 | |||
AK160248.1 | ||||
HQ438967.1 | ||||
HQ438968.1 | ||||
HQ438969.1 | ||||
HQ438970.1 | ||||
HQ438971.1 | ||||
HQ438972.1 | ||||
HQ438973.1 | ||||
HQ438974.1 | ||||
HQ438975.1 | ||||
HQ438976.1 | ||||
HQ438977.1 | ||||
HQ438978.1 | ||||
HQ438979.1 | ||||
HQ438980.1 | ||||
HQ438981.1 | ||||
HQ438982.1 | ||||
HQ438983.1 | ||||
HQ438984.1 | ||||
HQ438985.1 | ||||
HQ438986.1 | ||||
HQ438987.1 | ||||
HQ438988.1 | ||||
HQ438989.1 | ||||
HQ438990.1 | ||||
HQ438991.1 | ||||
HQ438992.1 | ||||
HQ438993.1 | ||||
HQ438994.1 | ||||
HQ438995.1 | ||||
HQ438996.1 | ||||
HQ438997.1 | ||||
HQ438998.1 | ||||
HQ438999.1 | ||||
HQ439000.1 | ||||
HQ439001.1 | ||||
HQ439002.1 | ||||
HQ439003.1 | ||||
HQ439004.1 | ||||
HQ439005.1 | ||||
HQ439006.1 | ||||
HQ439007.1 | ||||
HQ439008.1 | ||||
HQ439009.1 | ||||
HQ439010.1 | ||||
HQ439011.1 | ||||
HQ439012.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)